siRNA Details
Gene Symbol: FA2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024306.4 | NP_077282.3 | 7 | 2095 | ||
XM_011523319.2 | XP_011521621.1 | 7 | 1794 | |||
GenBank mRNA | AJ278219.1 | - | 6 | 1741 | ||
AK058016.1 | - | 7 | 2089 | |||
BC002679.2 | - | 7 | 2061 | 3610614 | ||
BC004263.1 | - | 7 | 2061 | 3611279 | ||
BC017049.1 | - | 7 | 2066 | 3850399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
