siRNA Details
Gene Symbol: GNPTAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI479646.1 | - | 3 | 410 | ||
BC002779.2 | - | 3 | 417 | |||
BC094884.1 | - | 3 | 417 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
