siRNA Details
Gene Symbol: TMEM243 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250589.3 | XP_005250646.1 | 5 | 819 | ||
XM_017012617.1 | XP_016868106.1 | 5 | 942 | |||
XM_017012618.1 | XP_016868107.1 | 5 | 686 | |||
GenBank mRNA | BC002837.2 | - | 4 | 603 | 3634983 | |
DB241717.1 | - | 4 | 490 |
