siRNA Details
Gene Symbol: PGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024419.4 | NP_077733.3 | 7 | 973 | ||
NR_110601.1 | 6 | 895 | ||||
NR_110602.1 | 6 | 857 | ||||
NR_111989.1 | 6 | 725 | ||||
XM_011525487.2 | XP_011523789.1 | 7 | 1348 | |||
XM_011525488.2 | XP_011523790.1 | 7 | 1348 | |||
XM_011525489.2 | XP_011523791.1 | 7 | 1348 | |||
XM_017025357.1 | XP_016880846.1 | 7 | 1348 | |||
XM_017025358.1 | XP_016880847.1 | 7 | 1348 | |||
XM_017025359.1 | XP_016880848.1 | 7 | 1348 | |||
XM_017025360.1 | XP_016880849.1 | 7 | 1348 | |||
XM_017025361.1 | XP_016880850.1 | 7 | 967 | |||
XM_017025362.1 | XP_016880851.1 | 7 | 967 | |||
XM_017025363.1 | XP_016880852.1 | 7 | 919 | |||
XM_017025364.1 | XP_016880853.1 | 6 | 869 | |||
XM_017025365.1 | XP_016880854.1 | 6 | 821 | |||
XM_017025366.1 | XP_016880855.1 | 7 | 967 | |||
GenBank mRNA | AK024529.1 | - | 6 | 693 | ||
AK225030.1 | - | 6 | 694 | |||
AK296882.1 | - | 7 | 947 | |||
AK297156.1 | - | 6 | 813 | |||
AK307874.1 | - | 7 | 949 | |||
AK309890.1 | - | 3 | 474 | |||
AK316147.1 | - | 7 | 951 | |||
AK316405.1 | - | 6 | 860 | |||
AL359590.1 | - | 2 | 70 | |||
BC008903.2 | - | 7 | 921 | |||
BC015570.2 | - | 7 | 921 | |||
BC025951.1 | - | 7 | 906 | |||
BC035662.1 | - | 6 | 856 | |||
BC108732.1 | - | 7 | 937 | 6504169 | ||
CR749720.1 | - | 2 | 70 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
