siRNA Details
Gene Symbol: CYP2R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005252789.2 | XP_005252846.1 | 1 | 609 | ||
XM_011519895.1 | XP_011518197.1 | 1 | 609 | |||
XM_017017190.1 | XP_016872679.1 | 1 | 609 | |||
XM_017017191.1 | XP_016872680.1 | 1 | 609 | |||
XM_017017192.1 | XP_016872681.1 | 1 | 609 | |||
XM_017017193.1 | XP_016872682.1 | 1 | 609 | |||
GenBank mRNA | BC084557.1 | - | 1 | 719 |
Gene Symbol: LOC107984314 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017018639.1 | XP_016874128.1 | 1 | 954 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
