siRNA Details
Gene Symbol: FOSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005253.3 | NP_005244.1 | 4 | 3495 | ||
XM_006711976.2 | XP_006712039.1 | 4 | 3699 | |||
XM_006711977.3 | XP_006712040.1 | 4 | 4552 | |||
XM_017003737.1 | XP_016859226.1 | 4 | 4208 | |||
GenBank mRNA | AK026959.1 | - | 1 | 1230 | ||
BC008899.2 | - | 1 | 1002 | |||
BC085197.1 | - | 1 | 1005 | |||
BX647822.1 | - | 4 | 2671 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
