siRNA Details
Gene Symbol: PRKCI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002740.5 | NP_002731.4 | 14, 15 | 1647 | ||
XM_017006860.1 | XP_016862349.1 | 13, 14 | 1554 | |||
GenBank mRNA | AB451443.1 | - | 14, 15 | 1382 | ||
AK315342.1 | - | 14, 15 | 1575 | |||
BC022016.2 | - | 14, 15 | 1642 | 4823853 | ||
BC042405.1 | - | 2, 3 | 413 | |||
L18964.1 | - | 14, 15 | 1646 | |||
L33881.1 | - | 14, 15 | 1586 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
