siRNA Details
Gene Symbol: ATP13A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024524.3 | NP_078800.3 | 11, 12 | 1560 | ||
XM_005269357.3 | XP_005269414.1 | 11, 12 | 1581 | |||
XM_011513120.2 | XP_011511422.1 | 12, 13 | 1682 | |||
XM_011513121.1 | XP_011511423.1 | 11, 12 | 1560 | |||
XM_011513122.1 | XP_011511424.1 | 11, 12 | 1254 | |||
XM_011513123.2 | XP_011511425.1 | 12, 13 | 1687 | |||
XM_011513124.2 | XP_011511426.1 | 12, 13 | 1687 | |||
GenBank mRNA | AJ306929.1 | - | 2, 3 | 2084 | ||
DR005713.1 | - | 4, 5 | 441 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
