siRNA Details
Gene Symbol: NES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006617.1 | NP_006608.1 | 4 | 3519 | ||
GenBank mRNA | AB073350.1 | - | 1 | 1578 | ||
AK025494.1 | - | 2 | 665 | |||
BC108285.1 | - | 1 | 154 | |||
BG912782.1 | - | 1 | 152 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
