siRNA Details
Gene Symbol: ATP13A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024524.3 | NP_078800.3 | 32 | 6119 | ||
XM_005269357.3 | XP_005269414.1 | 33 | 6230 | |||
XM_011513120.2 | XP_011511422.1 | 34 | 6331 | |||
XM_011513121.1 | XP_011511423.1 | 33 | 6209 | |||
XM_011513122.1 | XP_011511424.1 | 33 | 5903 | |||
XM_011513123.2 | XP_011511425.1 | 33 | 6246 | |||
GenBank mRNA | AJ306929.1 | - | 18 | 4017 | ||
BC037805.1 | - | 1 | 514 | |||
BC106085.1 | - | 5 | 2523 | |||
BX641098.1 | - | 2 | 3098 | |||
Z24724.1 | - | 1 | 665 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
