siRNA Details
Gene Symbol: CEP97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303401.1 | NP_001290330.1 | 11 | 4599 | ||
NM_024548.3 | NP_078824.2 | 11 | 4776 | |||
XM_006713743.3 | XP_006713806.1 | 10 | 5167 | |||
XM_011513127.1 | XP_011511429.1 | 11 | 4733 | |||
XM_017007178.1 | XP_016862667.1 | 10 | 4992 | |||
GenBank mRNA | AK026700.1 | - | 1 | 2549 | ||
AL833269.1 | - | 11 | 4726 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
