siRNA Details
Gene Symbol: NAA16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024561.4 | NP_078837.3 | 15, 16 | 2278 | ||
XM_005266523.2 | XP_005266580.1 | 8, 9 | 1130 | |||
XM_006719866.2 | XP_006719929.1 | 13, 14 | 2074 | |||
XM_011535227.2 | XP_011533529.1 | 15, 16 | 2254 | |||
XM_017020744.1 | XP_016876233.1 | 11, 12 | 1597 | |||
GenBank mRNA | AK025707.1 | - | 2, 3 | 316 | ||
AK291248.1 | - | 15, 16 | 2192 | |||
AL833341.1 | - | 9, 10 | 1255 | |||
BC028112.1 | - | 5, 6 | 865 | |||
BC101719.1 | - | 15, 16 | 2050 | 8069225 | ||
BC101721.1 | - | 15, 16 | 2050 | 8069227 | ||
BX640665.1 | - | 16, 17 | 2315 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
