siRNA Details
Gene Symbol: CCNJL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308173.1 | NP_001295102.1 | 6 | 2489 | ||
NM_024565.6 | NP_078841.3 | 7 | 2633 | |||
NR_131769.1 | 8 | 2698 | ||||
XM_006714917.3 | XP_006714980.1 | 6 | 3068 | |||
XM_011534646.2 | XP_011532948.1 | 6 | 2371 | |||
XM_011534647.2 | XP_011532949.1 | 4 | 2423 | |||
XM_017009846.1 | XP_016865335.1 | 7 | 2716 | |||
XM_017009847.1 | XP_016865336.1 | 5 | 2268 | |||
XM_017009848.1 | XP_016865337.1 | 5 | 2628 | |||
XM_017009849.1 | XP_016865338.1 | 4 | 2005 | |||
GenBank mRNA | AK024228.1 | - | 2 | 1608 | ||
AK131381.1 | - | 8 | 2698 | |||
BC013353.2 | - | 3 | 1669 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
