siRNA Details
Gene Symbol: C3orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171747.1 | NP_001165218.1 | 4 | 1568 | ||
NM_024616.2 | NP_078892.2 | 6 | 1821 | |||
XM_011513133.2 | XP_011511435.1 | 6 | 1730 | |||
XM_017007181.1 | XP_016862670.1 | 6 | 2795 | |||
GenBank mRNA | AK026839.1 | - | 6 | 1765 | ||
AY830714.1 | - | 6 | 1767 | |||
BC017064.1 | - | 6 | 1754 | 3885219 | ||
BX104180.1 | - | 1 | 268 |
