siRNA Details
Gene Symbol: QTRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256835.1 | NP_001243764.1 | 2 | 253 | ||
NM_024638.3 | NP_078914.1 | 3 | 361 | |||
XM_005247770.3 | XP_005247827.1 | 2 | 202 | |||
XM_011513137.2 | XP_011511439.1 | 2 | 224 | |||
XM_017007182.1 | XP_016862671.1 | 3 | 292 | |||
XM_017007184.1 | XP_016862673.1 | 3 | 292 | |||
GenBank mRNA | AK023022.1 | - | 3 | 251 | ||
AL833709.1 | - | 2 | 221 | |||
BC034559.2 | - | 2 | 235 | 4385906 | ||
BC039265.1 | - | 3 | 332 | 5296335 | ||
CR457296.1 | - | 1 | 102 | |||
DC400836.1 | - | 2 | 253 |
