siRNA Details
Gene Symbol: GALNT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024642.4 | NP_078918.3 | 2 | 418 | ||
XM_006717287.1 | XP_006717350.1 | 2 | 234 | |||
XM_011519018.2 | XP_011517320.1 | 3 | 1090 | |||
XM_011519020.1 | XP_011517322.1 | 2 | 234 | |||
XM_017015133.1 | XP_016870622.1 | 3 | 178 | |||
GenBank mRNA | AB040673.1 | - | 2 | 418 | ||
AB078146.1 | - | 2 | 418 | |||
AJ132365.2 | - | 2 | 418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
