siRNA Details
Gene Symbol: TBC1D17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168222.1 | NP_001161694.1 | 3 | 436 | ||
NM_024682.2 | NP_078958.2 | 4 | 535 | |||
XM_011527317.2 | XP_011525619.1 | 4 | 447 | |||
GenBank mRNA | AB449903.1 | - | 4 | 236 | ||
AK022230.1 | - | 4 | 294 | |||
AK090606.1 | - | 3 | 222 | |||
AK300007.1 | - | 3 | 195 | |||
AK309412.1 | - | 5 | 442 | |||
AY129013.1 | - | 2 | 983 | |||
BC003516.1 | - | 4 | 388 | 3050014 |
