siRNA Details
Gene Symbol: NFIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | HM204564.1 | - | 1 | 137 | ||
S79880.1 | - | 2 | 350 | |||
U26173.1 | - | 2 | 350 | |||
X64318.1 | - | 2 | 350 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
