siRNA Details
Gene Symbol: ZNF419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291743.1 | NP_001278672.1 | 2, 3 | 320 | ||
NM_024691.3 | NP_078967.3 | 2, 3 | 320 | |||
GenBank mRNA | BC008297.1 | - | 2, 3 | 266 | 3544044 | |
BC024259.1 | - | 2, 3 | 266 | 3604215 | ||
CT002615.1 | - | 2, 3 | 250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
