siRNA Details
Gene Symbol: PKN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002741.3 | NP_002732.3 | 6 | 938 | ||
NM_213560.1 | NP_998725.1 | 6 | 805 | |||
GenBank mRNA | AK123007.1 | - | 6 | 819 | ||
AK292130.1 | - | 6 | 802 | |||
AK313886.1 | - | 6 | 778 | |||
BC040061.1 | - | 6 | 935 | 5752583 | ||
BC094766.1 | - | 6 | 969 | 30342125 | ||
D26181.1 | - | 6 | 806 | |||
S75546.1 | - | 6 | 854 | |||
U33053.1 | - | 6 | 854 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
