siRNA Details
Gene Symbol: CPLX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030005.2 | NP_001025176.1 | 1 | 237 | ||
GenBank mRNA | AB072900.1 | - | 1 | 220 | ||
AK024055.1 | - | 1 | 237 | |||
AY286501.1 | - | 1 | 157 | |||
BC018026.1 | - | 1 | 220 | 4817723 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
