siRNA Details
Gene Symbol: BAHCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291324.1 | NP_001278253.1 | 28 | 10272 | ||
XM_011525063.2 | XP_011523365.1 | 29 | 10640 | |||
XM_017024898.1 | XP_016880387.1 | 28 | 10299 | |||
XM_017024899.1 | XP_016880388.1 | 27 | 10229 | |||
GenBank mRNA | AF086116.1 | - | 1 | 302 | ||
AK026711.1 | - | 1 | 1824 | |||
AK056636.1 | - | 1 | 2451 | |||
AL713748.1 | - | 3 | 2856 | |||
BC033222.1 | - | 3 | 2903 | |||
BC050673.2 | - | 1 | 1602 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
