siRNA Details
Gene Symbol: GIMAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024711.5 | NP_078987.3 | 2, 3 | 573 | ||
GenBank mRNA | AK300445.1 | - | 2, 3 | 289 | ||
AK308110.1 | - | 2, 3 | 238 | |||
AK314667.1 | - | 2, 3 | 277 | |||
AL541003.3 | - | 2, 3 | 280 | |||
BC060760.1 | - | 2, 3 | 234 | 30336063 | ||
BC074744.2 | - | 2, 3 | 123 | 30915204 | ||
CR749355.1 | - | 2, 3 | 310 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
