siRNA Details
Gene Symbol: QTRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256835.1 | NP_001243764.1 | 9 | 3818 | ||
NM_001256836.1 | NP_001243765.1 | 6 | 3464 | |||
NM_001256837.1 | NP_001243766.1 | 8 | 3562 | |||
NM_024638.3 | NP_078914.1 | 10 | 3926 | |||
XM_005247769.3 | XP_005247826.1 | 10 | 3707 | |||
XM_005247770.3 | XP_005247827.1 | 8 | 3690 | |||
XM_006713744.3 | XP_006713807.1 | 9 | 3559 | |||
XM_011513137.2 | XP_011511439.1 | 10 | 3830 | |||
XM_017007182.1 | XP_016862671.1 | 9 | 3780 | |||
XM_017007183.1 | XP_016862672.1 | 7 | 3545 | |||
XM_017007184.1 | XP_016862673.1 | 10 | 3821 | |||
GenBank mRNA | AK023022.1 | - | 10 | 3816 | ||
AL833709.1 | - | 9 | 3786 | |||
BC034559.2 | - | 9 | 3800 | 4385906 | ||
BC039265.1 | - | 10 | 3897 | 5296335 |
