siRNA Details
Gene Symbol: NOL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024654.4 | NP_078930.3 | 12 | 6396 | ||
GenBank mRNA | AK026976.1 | - | 9 | 2243 | ||
BC073860.1 | - | 9 | 2441 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
