siRNA Details
Gene Symbol: THAP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317776.1 | NP_001304705.1 | 6 | 2090 | ||
NM_024672.5 | NP_078948.3 | 5 | 2024 | |||
XM_005263220.2 | XP_005263277.1 | 5 | 1981 | |||
XM_011532246.2 | XP_011530548.1 | 5 | 1814 | |||
XM_017008603.1 | XP_016864092.1 | 5 | 2864 | |||
XM_017008604.1 | XP_016864093.1 | 5 | 2917 | |||
XM_017008605.1 | XP_016864094.1 | 5 | 1958 | |||
XM_017008606.1 | XP_016864095.1 | 6 | 2082 | |||
XM_017008607.1 | XP_016864096.1 | 5 | 2297 | |||
XM_017008608.1 | XP_016864097.1 | 6 | 2936 | |||
XM_017008609.1 | XP_016864098.1 | 6 | 2086 | |||
XM_017008610.1 | XP_016864099.1 | 6 | 2073 | |||
GenBank mRNA | AK026973.1 | - | 3 | 1354 | ||
AK091412.1 | - | 5 | 1921 | |||
AK225333.1 | - | 3 | 1361 | |||
BC131722.1 | - | 6 | 1971 | |||
BX648134.1 | - | 6 | 2244 |
