siRNA Details
Gene Symbol: TBC1D17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168222.1 | NP_001161694.1 | 8 | 1219 | ||
NM_024682.2 | NP_078958.2 | 9 | 1318 | |||
XM_011527317.2 | XP_011525619.1 | 9 | 1230 | |||
GenBank mRNA | AB449903.1 | - | 9 | 1019 | ||
AK022230.1 | - | 9 | 1077 | |||
AK090606.1 | - | 8 | 1019 | |||
AK300007.1 | - | 8 | 978 | |||
AY129013.1 | - | 7 | 1765 | |||
BC003516.1 | - | 9 | 1171 | 3050014 |
