siRNA Details
Gene Symbol: ZNF419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098491.1 | NP_001091961.1 | 5 | 853 | ||
NM_001098492.1 | NP_001091962.1 | 4 | 814 | |||
NM_001098493.1 | NP_001091963.1 | 4 | 811 | |||
NM_001098494.1 | NP_001091964.1 | 4 | 754 | |||
NM_001098495.1 | NP_001091965.1 | 3 | 715 | |||
NM_001098496.1 | NP_001091966.1 | 3 | 712 | |||
NM_001291743.1 | NP_001278672.1 | 4 | 751 | |||
NM_001291744.1 | NP_001278673.1 | 4 | 770 | |||
NM_024691.3 | NP_078967.3 | 5 | 850 | |||
GenBank mRNA | AB209371.1 | - | 3 | 4099 | ||
AK026886.1 | - | 4 | 724 | |||
AK302135.1 | - | 3 | 673 | |||
AK304569.1 | - | 4 | 759 | |||
AK307802.1 | - | 5 | 842 | |||
AK309427.1 | - | 3 | 1317 | |||
AK316156.1 | - | 4 | 772 | |||
AK316297.1 | - | 4 | 804 | |||
BC008297.1 | - | 5 | 796 | 3544044 | ||
BC024259.1 | - | 5 | 796 | 3604215 |
Gene Symbol: ZNF772 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024596.2 | NP_001019767.1 | 5 | 1060 | ||
NM_001144068.1 | NP_001137540.1 | 4 | 937 | |||
XM_005258943.4 | XP_005259000.1 | 3 | 4190 | |||
XM_005258944.3 | XP_005259001.1 | 2 | 4151 | |||
XM_011526980.2 | XP_011525282.1 | 5 | 4694 | |||
XM_017026813.1 | XP_016882302.1 | 4 | 4571 | |||
GenBank mRNA | AK293825.1 | - | 3 | 860 | ||
AK294941.1 | - | 3 | 765 | |||
AK301562.1 | - | 2 | 739 | |||
AK302574.1 | - | 4 | 899 | |||
BX647068.1 | - | 5 | 1060 | |||
CR749225.1 | - | 2 | 746 |
Gene Symbol: ZNF773 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304334.1 | NP_001291263.1 | 4 | 798 | ||
NM_001304336.1 | NP_001291265.1 | 4 | 794 | |||
NM_198542.2 | NP_940944.1 | 4 | 801 | |||
GenBank mRNA | AK057209.1 | - | 4 | 717 | ||
BC005167.2 | - | 4 | 714 | 3535599 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
