siRNA Details
Gene Symbol: ECHDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024693.4 | NP_078969.2 | 5 | 1308 | ||
XM_011519689.1 | XP_011517991.1 | 4 | 1082 | |||
GenBank mRNA | AF289604.1 | - | 6 | 1382 | ||
AK024562.1 | - | 5 | 1220 | |||
AK222511.1 | - | 5 | 1247 | |||
AK290902.1 | - | 5 | 1238 | |||
BC001091.1 | - | 5 | 1041 | |||
BC009617.1 | - | 5 | 1230 | 3893103 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
