siRNA Details
Gene Symbol: ADGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024694.3 | NP_078970.3 | 36 | 5039 | ||
XM_006715566.3 | XP_006715629.1 | 36 | 5074 | |||
XM_011536125.2 | XP_011534427.1 | 36 | 5025 | |||
XM_011536128.2 | XP_011534430.1 | 35 | 4897 | |||
XM_011536130.2 | XP_011534432.1 | 37 | 4973 | |||
XM_011536131.2 | XP_011534433.1 | 30 | 4247 | |||
XM_011536132.2 | XP_011534434.1 | 28 | 3835 | |||
XM_011536134.2 | XP_011534436.1 | 32 | 6528 | |||
XM_011536135.2 | XP_011534437.1 | 18 | 2716 | |||
XM_017011314.1 | XP_016866803.1 | 36 | 5071 | |||
XM_017011315.1 | XP_016866804.1 | 36 | 5023 | |||
GenBank mRNA | AK026774.1 | - | 12 | 1872 | ||
AL832192.1 | - | 32 | 6530 | |||
BG199812.1 | - | 1 | 169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
