siRNA Details
Gene Symbol: BAHCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291324.1 | NP_001278253.1 | 28 | 10365 | ||
XM_011525063.2 | XP_011523365.1 | 29 | 10733 | |||
XM_017024898.1 | XP_016880387.1 | 28 | 10392 | |||
XM_017024899.1 | XP_016880388.1 | 27 | 10322 | |||
GenBank mRNA | AF086116.1 | - | 1 | 395 | ||
AK026711.1 | - | 1 | 1917 | |||
AL713748.1 | - | 3 | 2949 | |||
BC033222.1 | - | 3 | 2996 | |||
BC050673.2 | - | 1 | 1695 |

More Information
Other Species Targeted

Gene Ontology Categories:
