siRNA Details
Gene Symbol: ZNF385D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024697.2 | NP_078973.1 | 6 | 1173 | ||
XM_017007191.1 | XP_016862680.1 | 8 | 1211 | |||
XM_017007192.1 | XP_016862681.1 | 7 | 1154 | |||
XM_017007193.1 | XP_016862682.1 | 8 | 1201 | |||
XM_017007194.1 | XP_016862683.1 | 9 | 1266 | |||
XM_017007195.1 | XP_016862684.1 | 7 | 906 | |||
XM_017007196.1 | XP_016862685.1 | 7 | 1384 | |||
XM_017007197.1 | XP_016862686.1 | 7 | 1622 | |||
XM_017007198.1 | XP_016862687.1 | 7 | 1377 | |||
XM_017007199.1 | XP_016862688.1 | 6 | 732 | |||
XM_017007200.1 | XP_016862689.1 | 6 | 2948 | |||
XM_017007201.1 | XP_016862690.1 | 6 | 753 | |||
XM_017007202.1 | XP_016862691.1 | 4 | 469 | |||
GenBank mRNA | AK026072.1 | - | 6 | 1173 | ||
BC007212.1 | - | 6 | 997 | 3347783 |
