siRNA Details
Gene Symbol: DHRS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270424.1 | NP_001257353.1 | 10 | 1149 | ||
NM_024705.2 | NP_078981.1 | 8 | 1925 | |||
XM_005266527.3 | XP_005266584.1 | 9 | 1072 | |||
XM_017020754.1 | XP_016876243.1 | 8 | 1006 | |||
GenBank mRNA | AK023701.1 | - | 8 | 1920 | ||
BC026024.1 | - | 10 | 1082 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
