siRNA Details
Gene Symbol: ASB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024708.3 | NP_078984.2 | 5 | 1649 | ||
GenBank mRNA | AK026204.1 | - | 5 | 1569 | ||
AK222958.1 | - | 5 | 1569 | |||
AK222974.1 | - | 5 | 1569 | |||
BC063581.1 | - | 5 | 1590 | 4370796 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
