siRNA Details
Gene Symbol: AGMAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024758.4 | NP_079034.3 | 2 | 580 | ||
GenBank mRNA | AK130034.1 | - | 2 | 376 | ||
AY057097.1 | - | 2 | 480 | |||
BC005090.1 | - | 2 | 580 | 3837251 | ||
BM788254.1 | - | 2 | 511 | |||
DN996958.1 | - | 2 | 570 |
