siRNA Details
Gene Symbol: TM4SF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024795.3 | NP_079071.2 | 1 | 154 | ||
GenBank mRNA | AK026453.1 | - | 1 | 153 | ||
AY358671.1 | - | 1 | 146 | |||
BC035754.1 | - | 1 | 141 | 5760060 | ||
BC137256.1 | - | 1 | 140 | 9021253 | ||
BC137257.1 | - | 1 | 140 | 9021254 | ||
DA926038.1 | - | 1 | 154 |
