siRNA Details
Gene Symbol: BAALC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024812.2 | NP_079088.1 | 2, 3 | 540 | ||
GenBank mRNA | AF371319.1 | - | 2, 3 | 335 | ||
AK022077.1 | - | 2, 3 | 518 | |||
BC011517.1 | - | 2, 3 | 482 | 4183341 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
