siRNA Details
Gene Symbol: PRKD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002742.2 | NP_002733.2 | 2 | 561 | ||
XM_005267859.1 | XP_005267916.1 | 2 | 495 | |||
XM_011536964.2 | XP_011535266.1 | 2 | 2917 | |||
XM_011536965.2 | XP_011535267.1 | 2 | 4967 | |||
XM_017021461.1 | XP_016876950.1 | 2 | 272 | |||
XM_017021462.1 | XP_016876951.1 | 2 | 182 | |||
GenBank mRNA | AK314170.1 | - | 2 | 497 | ||
CN412379.1 | - | 2 | 149 | |||
X75756.1 | - | 2 | 615 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
