siRNA Details
Gene Symbol: ZNF672 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024836.2 | NP_079112.1 | 4 | 2198 | ||
XM_005270336.1 | XP_005270393.1 | 4 | 2043 | |||
GenBank mRNA | AK025954.1 | - | 4 | 2045 | ||
AK027476.1 | - | 1 | 1223 | |||
AK027679.1 | - | 2 | 1592 | |||
AK298016.1 | - | 4 | 1285 | |||
BC007385.2 | - | 1 | 652 | |||
BC035140.1 | - | 1 | 2578 | 5264126 | ||
BC050348.1 | - | 1 | 2574 | 5260682 | ||
BC068506.1 | - | 4 | 1896 | 4824147 |
