siRNA Details
Gene Symbol: CLMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024734.3 | NP_079010.2 | 10 | 2719 | ||
XM_011537158.1 | XP_011535460.1 | 10 | 2715 | |||
XM_011537159.2 | XP_011535461.1 | 10 | 2715 | |||
XM_017021646.1 | XP_016877135.1 | 10 | 2885 | |||
XM_017021647.1 | XP_016877136.1 | 10 | 2715 | |||
GenBank mRNA | AB047979.1 | - | 10 | 2676 | ||
AK023828.1 | - | 2 | 1685 | |||
AK314315.1 | - | 10 | 2717 | |||
BC014554.2 | - | 2 | 1259 | |||
BC025251.1 | - | 2 | 1658 | |||
BC068482.1 | - | 10 | 2668 | 4375469 |
