siRNA Details
Gene Symbol: WDR78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024763.4 | NP_079039.4 | 12, 13 | 1940 | ||
XM_005271212.3 | XP_005271269.1 | 11, 12 | 1698 | |||
XM_011542161.2 | XP_011540463.1 | 11, 12 | 1806 | |||
XM_011542162.2 | XP_011540464.1 | 12, 13 | 1893 | |||
XM_017002353.1 | XP_016857842.1 | 11, 12 | 1698 | |||
XM_017002354.1 | XP_016857843.1 | 10, 11 | 1611 | |||
XM_017002355.1 | XP_016857844.1 | 9, 10 | 1313 | |||
XM_017002356.1 | XP_016857845.1 | 10, 11 | 1557 | |||
GenBank mRNA | AK026782.1 | - | 8, 9 | 1181 | ||
AK127011.1 | - | 9, 10 | 2309 | |||
AK292830.1 | - | 13, 14 | 1893 | |||
AL133617.1 | - | 3, 4 | 336 | |||
BC126379.1 | - | 12, 13 | 1915 | 8992095 | ||
BC126381.1 | - | 12, 13 | 1915 | 8992097 | ||
BX648840.1 | - | 12, 13 | 1940 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
