siRNA Details
Gene Symbol: ZNF669 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142572.1 | NP_001136044.1 | 4 | 1301 | ||
NM_024804.2 | NP_079080.2 | 4 | 1559 | |||
XM_005273270.2 | XP_005273327.1 | 3 | 1268 | |||
XM_005273271.2 | XP_005273328.1 | 3 | 1526 | |||
GenBank mRNA | AK022668.1 | - | 4 | 1513 | ||
AK056020.1 | - | 4 | 1290 | |||
BC015312.1 | - | 4 | 1559 | 4413005 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
