siRNA Details
Gene Symbol: C11orf63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024806.3 | NP_079082.2 | 5 | 2158 | ||
XM_005271680.4 | XP_005271737.1 | 5 | 2019 | |||
XM_006718915.2 | XP_006718978.1 | 4 | 1779 | |||
XM_011543009.2 | XP_011541311.1 | 5 | 2227 | |||
XM_017018338.1 | XP_016873827.1 | 5 | 1893 | |||
XM_017018339.1 | XP_016873828.1 | 5 | 1926 | |||
XM_017018340.1 | XP_016873829.1 | 5 | 1737 | |||
XM_017018341.1 | XP_016873830.1 | 5 | 2508 | |||
XM_017018342.1 | XP_016873831.1 | 5 | 1893 | |||
XM_017018343.1 | XP_016873832.1 | 4 | 1779 | |||
XM_017018344.1 | XP_016873833.1 | 2 | 748 | |||
GenBank mRNA | AK027207.1 | - | 4 | 1512 | ||
AK291625.1 | - | 5 | 1761 | |||
BC068507.1 | - | 5 | 1921 | 5298472 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
