siRNA Details
Gene Symbol: RABEP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024816.2 | NP_079092.2 | 13 | 2240 | ||
GenBank mRNA | AK026935.1 | - | 13 | 2239 | ||
AK092299.1 | - | 13 | 2781 | |||
BC046118.1 | - | 13 | 2234 | |||
BC058900.1 | - | 11 | 2135 | 5415624 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
