siRNA Details
Gene Symbol: SPEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024867.3 | NP_079143.3 | 25 | 3668 | ||
XM_005248376.3 | XP_005248433.1 | 25 | 3773 | |||
XM_005248377.3 | XP_005248434.1 | 25 | 3758 | |||
XM_011514135.2 | XP_011512437.1 | 25 | 3778 | |||
XM_011514136.2 | XP_011512438.1 | 25 | 3777 | |||
XM_011514137.2 | XP_011512439.1 | 25 | 3762 | |||
XM_011514138.2 | XP_011512440.1 | 25 | 3754 | |||
XM_011514139.2 | XP_011512441.1 | 24 | 3702 | |||
XM_011514140.2 | XP_011512442.1 | 24 | 3512 | |||
XM_011514141.2 | XP_011512443.1 | 26 | 3976 | |||
XM_011514142.1 | XP_011512444.1 | 25 | 3749 | |||
XM_017009880.1 | XP_016865369.1 | 24 | 3594 | |||
XM_017009881.1 | XP_016865370.1 | 25 | 4632 | |||
GenBank mRNA | AB051557.1 | - | 15 | 2009 | ||
AK027230.1 | - | 2 | 114 | |||
AK225586.1 | - | 2 | 114 | |||
AK225607.1 | - | 2 | 114 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
