siRNA Details
Gene Symbol: LINC00472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026807.2 | 2 | 695 | |||
NR_121612.1 | 2 | 695 | ||||
GenBank mRNA | AI808466.1 | - | 2 | 167 | ||
AI985403.1 | - | 2 | 217 | |||
AK023251.1 | - | 2 | 695 | |||
AW274058.1 | - | 2 | 201 |
