siRNA Details
Gene Symbol: RIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319987.1 | NP_001306916.1 | 2, 3 | 261 | ||
NM_024832.4 | NP_079108.3 | 3, 4 | 514 | |||
XM_011537164.2 | XP_011535466.1 | 2, 3 | 249 | |||
XM_017021651.1 | XP_016877140.1 | 3, 4 | 396 | |||
XM_017021652.1 | XP_016877141.1 | 3, 4 | 522 | |||
XM_017021653.1 | XP_016877142.1 | 3, 4 | 522 | |||
XM_017021654.1 | XP_016877143.1 | 3, 4 | 522 | |||
GenBank mRNA | AB081753.1 | - | 3, 4 | 362 | ||
AK021762.1 | - | 3, 4 | 514 | |||
AK128338.1 | - | 2, 3 | 261 | |||
BC070062.1 | - | 3, 4 | 502 | 30332851 | ||
BX248258.1 | - | 3, 4 | 500 | |||
DR003779.1 | - | 3, 4 | 554 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
