siRNA Details
Gene Symbol: ZNF746 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163474.1 | NP_001156946.1 | 4 | 708 | ||
NM_152557.4 | NP_689770.3 | 4 | 708 | |||
XM_005249955.2 | XP_005250012.1 | 4 | 778 | |||
XM_005249956.2 | XP_005250013.1 | 4 | 778 | |||
XM_005249957.1 | XP_005250014.1 | 4 | 880 | |||
XM_011515838.1 | XP_011514140.1 | 4 | 835 | |||
GenBank mRNA | AK291814.1 | - | 4 | 644 | ||
BC068505.1 | - | 4 | 604 | 30347892 |
Gene Symbol: ZNF767P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027788.1 | 3 | 580 | |||
NR_027789.1 | 4 | 707 | ||||
GenBank mRNA | AK022762.1 | - | 3 | 517 | ||
AK295771.1 | - | 4 | 545 | |||
BC047675.1 | - | 4 | 669 | 5727587 | ||
DA418417.1 | - | 3 | 491 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
