siRNA Details
Gene Symbol: PCNX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014801.3 | NP_055616.3 | 10 | 2647 | ||
XM_006711816.3 | XP_006711879.1 | 10 | 2469 | |||
XM_011544278.2 | XP_011542580.2 | 3 | 258 | |||
XM_017002391.1 | XP_016857880.1 | 9 | 2211 | |||
GenBank mRNA | AK021445.1 | - | 4 | 623 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
