siRNA Details
Gene Symbol: TRMT44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK022953.1 | - | 1 | 466 | ||
AK092387.1 | - | 2 | 2404 | |||
AK096417.1 | - | 1 | 1783 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
